ENA388699822 Enamine

6-bromo-2,2-dimethyl-3,4-dihydro-2H-1-benzopyran

  • MDL number MFCD11870667
  • Cas Number : 174894-80-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-123081

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-123081
Availability details for
Quantity
  • Availability
  • Add to Cart