ENA388699824 Enamine

2-(bromomethyl)-6-(propan-2-yl)pyridine

  • MDL number MFCD18259890
  • Cas Number : 442910-36-3
  • Molecular Weight : 214.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2-(bromomethyl)-6-isopropylpyridine
Inchi Code 1S/C9H12BrN/c1-7(2)9-5-3-4-8(6-10)11-9/h3-5,7H,6H2,1-2H3
Inchi Key DNFLPQFSDKFUCX-UHFFFAOYSA-N

Description

Purity 95
Storage temp. +4
Physical Form liquid
Shipping Temperature Ice Pack
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-123086

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-123086
Availability details for
Quantity
  • Availability
  • Add to Cart