ENA388699825 Enamine

6-chloro-1,3-dihydro-2$l^{6},1-benzothiazole-2,2-dione

  • MDL number MFCD19105166
  • Cas Number : 111248-91-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 149...151
Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-123087

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-123087
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart