ENA388699839 Enamine

2-amino-3-cyclopentylpropan-1-ol

  • MDL number MFCD19216186
  • Cas Number : 98486-67-0
  • Molecular Weight : 143.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form liquid
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-123104

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-123104
Availability details for
Quantity
  • Availability
  • Add to Cart