ENA388699842 Enamine

5-methyl-1-(oxan-4-yl)-1H-pyrazole-4-carboxylic acid

  • MDL number MFCD23144052
  • Cas Number : 1424188-16-8
  • Molecular Weight : 210.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 5-methyl-1-tetrahydro-2H-pyran-4-yl-1H-pyrazole-4-carboxylic acid
Inchi Code 1S/C10H14N2O3/c1-7-9(10(13)14)6-11-12(7)8-2-4-15-5-3-8/h6,8H,2-5H2,1H3,(H,13,14)
Inchi Key KURHRJDWCKRYDJ-UHFFFAOYSA-N

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-123109

Safety Information

signalword Warning
pcodes P261,P264,P270,P271,P280,P301+P312,P302+P352,P304+P340,P305+P351+P338,P312,P330,P332+P313,P337+P313,P362,P403+P233,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-123109
pictograms GHS07
hstatements H302,H315,H319,H335
Availability details for
Quantity
  • Availability
  • Add to Cart