ENA388699846 Enamine

2-[4-(2-phenoxyethoxy)phenyl]acetic acid

  • MDL number MFCD02664763
  • Cas Number : 383134-87-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 169...171
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-123113

Safety Information

msds https://www.enaminestore.com/catalog/EN300-123113
Availability details for
Quantity
  • Availability
  • Add to Cart