ENA388699851 Enamine

(9H-fluoren-9-yl)methyl N-[4-(hydroxymethyl)phenyl]carbamate

  • MDL number MFCD04974272
  • Cas Number : 475160-83-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 184-185
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-123119

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-123119
Availability details for
Quantity
  • Availability
  • Add to Cart