ENA388699854 Enamine

methyl 2-(2-methylpropane-2-sulfonyl)acetate

  • MDL number MFCD12171519
  • Cas Number : 63864-29-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-123123

Safety Information

msds https://www.enaminestore.com/catalog/EN300-123123
Availability details for
Quantity
  • Availability
  • Add to Cart