ENA388699856 Enamine

4-(bromomethyl)-3-methoxybenzonitrile

  • MDL number MFCD07368343
  • Cas Number : 104436-60-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 89...91
Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-123137

Safety Information

msds https://www.enaminestore.com/catalog/EN300-123137
Availability details for
Quantity
  • Availability
  • Add to Cart