ENA388699857 Enamine

1-(2-methoxyethyl)-2-propylpiperazine

  • MDL number MFCD20427454
  • Cas Number : 1343711-61-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-123138

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-123138
Availability details for
Quantity
  • Availability
  • Add to Cart