ENA388699863 Enamine

2-(4-chlorophenyl)-2-hydroxypropanoic acid

  • MDL number MFCD11052509
  • Cas Number : 4445-13-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 136-137
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-123145

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-123145
Availability details for
Quantity
  • Availability
  • Add to Cart