ENA388699866 Enamine

ethyl 3,6-dimethylpyridazine-4-carboxylate

  • MDL number MFCD23144057
  • Cas Number : 897008-37-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 49...51
Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-123148

Safety Information

msds https://www.enaminestore.com/catalog/EN300-123148
Availability details for
Quantity
  • Availability
  • Add to Cart