ENA388699868 Enamine

1-(3-bromopyridin-2-yl)-1H-pyrazol-4-amine

  • MDL number MFCD14651584
  • Cas Number : 1247866-28-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-123150

Safety Information

msds https://www.enaminestore.com/catalog/EN300-123150
Availability details for
Quantity
  • Availability
  • Add to Cart