ENA388699869 Enamine

(1R)-1-(4-fluoro-3-methylphenyl)propan-1-ol

  • MDL number MFCD15478225
  • Cas Number : 1176589-67-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-123151

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-123151
Availability details for
Quantity
  • Availability
  • Add to Cart