ENA388699872 Enamine

ethyl 5-(4-methyl-1,3-thiazol-5-yl)-1,3-oxazole-4-carboxylate

  • MDL number MFCD16065678
  • Cas Number : 1249854-25-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-123154

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-123154
Availability details for
Quantity
  • Availability
  • Add to Cart