ENA388699873 Enamine

methyl 2-(3-bromophenyl)-2-(methylamino)acetate hydrochloride

  • MDL number MFCD23144061
  • Cas Number : 1423358-94-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 173...175
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-123155

Safety Information

msds https://www.enaminestore.com/catalog/EN300-123155
Availability details for
Quantity
  • Availability
  • Add to Cart