ENA388699875 Enamine

1,1,3,3-tetramethyl-2,3-dihydro-1H-inden-5-ol

  • MDL number MFCD18970701
  • Cas Number : 53718-26-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-123157

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-123157
Availability details for
Quantity
  • Availability
  • Add to Cart