ENA388699886 Enamine

{3-[(methylsulfanyl)methyl]phenyl}boronic acid

  • MDL number MFCD14687401
  • Cas Number : 1333345-25-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-123189

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-123189
Availability details for
Quantity
  • Availability
  • Add to Cart