ENA388699887 Enamine

cyclopropyl(4-methoxy-2-methylphenyl)methanol

  • MDL number MFCD12787094
  • Cas Number : 1249951-65-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 39...41
Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-123190

Safety Information

msds https://www.enaminestore.com/catalog/EN300-123190
Availability details for
Quantity
  • Availability
  • Add to Cart