ENA388699891 Enamine

2-(5-chloro-1-methyl-1H-1,3-benzodiazol-2-yl)-6-methylphenol

  • MDL number MFCD12720913
  • Cas Number : 1178510-11-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-123195

Safety Information

msds https://www.enaminestore.com/catalog/EN300-123195
Availability details for
Quantity
  • Availability
  • Add to Cart