ENA388699892 Enamine

[2-methyl-6-(propan-2-yl)imidazo[2,1-b][1,3,4]thiadiazol-5-yl]methanol

  • MDL number MFCD19279063
  • Cas Number : 1267798-65-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-123196

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-123196
Availability details for
Quantity
  • Availability
  • Add to Cart