ENA388699893 Enamine

(1S)-1-(4-aminophenyl)ethan-1-ol

  • MDL number MFCD19204822
  • Cas Number : 500229-84-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-123197

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-123197
Availability details for
Quantity
  • Availability
  • Add to Cart