ENA388699900 Enamine

3-(methylsulfamoyl)propanoic acid

  • MDL number MFCD11168961
  • Cas Number : 933714-87-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-123205

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-123205
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart