ENA388699901 Enamine

1-(1,3-thiazol-4-yl)ethan-1-amine

  • MDL number MFCD06738792
  • Cas Number : 885279-02-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-123206

Safety Information

msds https://www.enaminestore.com/catalog/EN300-123206
Availability details for
Quantity
  • Availability
  • Add to Cart