ENA388699906 Enamine

2-(1-methoxy-2-methylpropylidene)propanedinitrile

  • MDL number MFCD09756476
  • Cas Number : 54122-57-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-123212

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-123212
Availability details for
Quantity
  • Availability
  • Add to Cart