ENA388699909 Enamine

3-methoxy-3-methylazetidine hydrochloride

  • MDL number MFCD11100995
  • Cas Number : 905843-93-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. 0
Shipping Temperature Ice Pack
Product Link https://www.enaminestore.com/catalog/EN300-123215

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-123215
Availability details for
Quantity
  • Availability
  • Add to Cart