ENA388699911 Enamine

3-(2-oxo-1,2-dihydroquinolin-3-yl)propanoic acid

  • MDL number MFCD09895305
  • Cas Number : 37618-03-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 227...229
Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-118139

Safety Information

msds https://www.enaminestore.com/catalog/EN300-118139
Availability details for
Quantity
  • Availability
  • Add to Cart