ENA388699916 Enamine

3-[(1H-indazol-4-yl)sulfamoyl]thiophene-2-carboxylic acid

  • MDL number MFCD23144072
  • Cas Number : 1178919-92-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-123275

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-123275
Availability details for
Quantity
  • Availability
  • Add to Cart