ENA388699923 Enamine

N-(3-bromophenyl)pyridine-3-carboxamide

  • MDL number MFCD01213560
  • Cas Number : 332174-80-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-123282

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-123282
Availability details for
Quantity
  • Availability
  • Add to Cart