ENA388699928 Enamine

3-methyl-[1,2]oxazolo[5,4-d]pyrimidin-4-amine

  • MDL number MFCD13813767
  • Cas Number : 89799-07-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 269...271
Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-123288

Safety Information

msds https://www.enaminestore.com/catalog/EN300-123288
Availability details for
Quantity
  • Availability
  • Add to Cart