ENA388699929 Enamine

4-methyl-3-(pyrrolidin-2-yl)-4H-1,2,4-triazole

  • MDL number MFCD16850766
  • Cas Number : 1247079-34-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-123289

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-123289
Availability details for
Quantity
  • Availability
  • Add to Cart