ENA388699930 Enamine

4-(3-hydroxyphenoxy)-N-methylpyridine-2-carboxamide

  • MDL number MFCD13159492
  • Cas Number : 1183073-05-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 124...126
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-123290

Safety Information

msds https://www.enaminestore.com/catalog/EN300-123290
Availability details for
Quantity
  • Availability
  • Add to Cart