ENA388699931 Enamine

1H-indazole-3,5-diamine

  • MDL number MFCD13813188
  • Cas Number : 19335-14-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 178-182
Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-118206

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-118206
Availability details for
Quantity
  • Availability
  • Add to Cart