ENA388699933 Enamine

4-chloro-3-ethyl-1H-pyrazole-5-carboxylic acid

  • MDL number MFCD16103389
  • Cas Number : 158668-22-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 199...201
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-123294

Safety Information

msds https://www.enaminestore.com/catalog/EN300-123294
Availability details for
Quantity
  • Availability
  • Add to Cart