ENA388699936 Enamine

3-(2-methylcyclopropyl)-3-oxopropanenitrile

  • MDL number MFCD10693826
  • Cas Number : 1152599-39-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. -10
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-123299

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-123299
Availability details for
Quantity
  • Availability
  • Add to Cart