ENA388699938 Enamine

(1R)-1-(3-fluoro-4-methylphenyl)propan-1-ol

  • MDL number MFCD15478223
  • Cas Number : 1270296-17-7
  • Molecular Weight : 168.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (R)-1-(3-fluoro-4-methylphenyl)propan-1-ol
Inchi Code 1S/C10H13FO/c1-3-10(12)8-5-4-7(2)9(11)6-8/h4-6,10,12H,3H2,1-2H3/t10-/m1/s1
Inchi Key ZZCGKYXNTIQVJM-SNVBAGLBSA-N

Description

Purity 95
Storage temp. +4
Physical Form liquid
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-123301

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-123301
Availability details for
Quantity
  • Availability
  • Add to Cart