ENA388699939 Enamine

1-(2,2-dimethylpiperazin-1-yl)ethan-1-one

  • MDL number MFCD19285183
  • Cas Number : 1266872-99-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-123302

Safety Information

msds https://www.enaminestore.com/catalog/EN300-123302
Availability details for
Quantity
  • Availability
  • Add to Cart