ENA388699942 Enamine

(2,6-dichlorophenyl)(oxan-4-yl)methanol

  • MDL number MFCD19611691
  • Cas Number : 1341787-90-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-123305

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-123305
Availability details for
Quantity
  • Availability
  • Add to Cart