ENA388699944 Enamine

N-[2-(4-chlorophenyl)ethyl]-6-methylpyridazin-3-amine

  • MDL number MFCD13477469
  • Cas Number : 1304082-94-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-123307

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-123307
Availability details for
Quantity
  • Availability
  • Add to Cart