ENA388699946 Enamine

tert-butyl 4-(sulfanylmethyl)piperidine-1-carboxylate

  • MDL number MFCD19689418
  • Cas Number : 581060-27-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. -20
Shipping Temperature Ice Pack
Product Link https://www.enaminestore.com/catalog/EN300-123309

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-123309
Availability details for
Quantity
  • Availability
  • Add to Cart