ENA388699948 Enamine

2-[(piperazin-1-yl)methyl]pyrazine

  • MDL number MFCD04973504
  • Cas Number : 1083300-35-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-123311

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-123311
Availability details for
Quantity
  • Availability
  • Add to Cart