ENA388699949 Enamine

7-(4-fluorophenoxy)-2,3-dihydro-1,4-benzodioxin-6-amine

  • MDL number MFCD12446418
  • Cas Number : 1019509-34-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-123313

Safety Information

msds https://www.enaminestore.com/catalog/EN300-123313
Availability details for
Quantity
  • Availability
  • Add to Cart