ENA388699954 Enamine

3-(1,3-dioxo-2,3-dihydro-1H-isoindol-2-yl)propane-1-sulfonamide

  • MDL number MFCD23144076
  • Cas Number : 6712-89-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 173...175
Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-123321

Safety Information

msds https://www.enaminestore.com/catalog/EN300-123321
Availability details for
Quantity
  • Availability
  • Add to Cart