ENA388699958 Enamine

2-bromo-6-chloro-4-fluorobenzene-1-sulfonyl chloride

  • MDL number MFCD12026141
  • Cas Number : 1208076-43-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 55...57
Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-123326

Safety Information

msds https://www.enaminestore.com/catalog/EN300-123326
Availability details for
Quantity
  • Availability
  • Add to Cart