ENA388699963 Enamine

ethyl 3-oxo-5-phenylpentanoate

  • MDL number MFCD00041348
  • Cas Number : 17071-29-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-123350

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-123350
Availability details for
Quantity
  • Availability
  • Add to Cart