ENA388699964 Enamine

(4-methoxy-2-methylphenyl)(4-methoxyphenyl)methanol

  • MDL number MFCD16357385
  • Cas Number : 36801-17-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 71...73
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-123351

Safety Information

msds https://www.enaminestore.com/catalog/EN300-123351
Availability details for
Quantity
  • Availability
  • Add to Cart