ENA388699978 Enamine

3-[2-(trifluoromethyl)phenyl]butan-2-amine

  • MDL number MFCD18288800
  • Cas Number : 21894-66-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-123370

Safety Information

msds https://www.enaminestore.com/catalog/EN300-123370
Availability details for
Quantity
  • Availability
  • Add to Cart