ENA388699989 Enamine

2-amino-4-phenyl-4,5-dihydro-1H-imidazol-5-one

  • MDL number MFCD20697368
  • Cas Number : 619330-78-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-119238

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-119238
Availability details for
Quantity
  • Availability
  • Add to Cart