ENA388699990 Enamine

3-amino-3-phenyl-N-[1-(pyridin-4-yl)ethyl]propanamide dihydrochloride

  • MDL number MFCD23144088
  • Cas Number : 1424350-44-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-123386

Safety Information

msds https://www.enaminestore.com/catalog/EN300-123386
Availability details for
Quantity
  • Availability
  • Add to Cart