ENA388699991 Enamine

1-[4-(propan-2-yl)phenyl]propan-2-amine

  • MDL number MFCD08452703
  • Cas Number : 32560-62-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-123387

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-123387
Availability details for
Quantity
  • Availability
  • Add to Cart